missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Sheep IFNG / Interferon Gamma Protein

A DNA sequence encoding the Ovis aries (Sheep) IFNG / Interferon Gamma Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6206-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16081550

  • 402.00 € / 10µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001009803.1
IFNG / Interferon Gamma Protein
Research Use Only
IFNG
Recombinant Protein
E. coli
QGPFFKEIEN LKEYFNASNP DVAKGGPLFS EILKNWKEES DKKIIQSQIV SFYFKLFENL KDNQVIQRSM DIIKQDMFQK FLNGSSEKLE DFKRLIQIPV DDLQIQRKAI NELIKVMNDL SPKSNLRKRK RSQNLFRGRR ASM
Greater than 90% as determined by SDS-PAGE.
443396
10 μg
Not determined.
Full Length
Sheep
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers