missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant SARS-CoV-2 NSP8 His-tag Protein
Brand: R&D Systems™ 10633-CV-100
This item is not returnable.
View return policy
Specifications
YP_009725304.1 | |
100 μg | |
COVID-19 | |
Supplied as a 0.2 μm filtered solution in HEPES and NaCl. |
Unconjugated | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
SARS-CoV-2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction