missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus fabD Protein

A DNA sequence encoding the Staphylococcus aureus (strain COL) fabD, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7663-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16070051

  • 920.10 € / 100µg

Estimated Shipment: 10-01-2022
Add to basket

Description

Description

Specifications

Specifications

WP_000047348.1
Greater than 90% as determined by SDS-PAGE.
Research Use Only
fabD
Recombinant Protein
E. coli
MSKTAIIFPG QGAQKVGMAQ DLFNNNDQAT EILTSAANTL DFDILETMFT DEEGKLGETE NTQPALLTHS SALLAALKNL NPDFTMGHSL GEYSSLVAAD VLSFEDAVKI VRKRGQLMAQ AFPTGVGSMA AVLGLDFDKV DEICKSLSSD DKIIEPANIN CPGQIVVSGH KALIDELVEK GKSLGAKRVM PLAVSGPFHS SLMKVIEEDF SSYINQFEWR DAKFPVVQNV NAQGETDKEV IKSNMVKQLY SPVQFINSTE WLIDQGVDHF IEIGPGKVLS GLIKKINRDV KLTSIQTLED VKGWNEND
S. aureus fabD Protein
100 μg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents
Certificates
Promotions

Promotions