missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Enterotoxin type E Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Pyruvate carboxylase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7046-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16037170

  • 3375.00 € / 1mg

Estimated Shipment: 22-11-2022
to see stock.
Add to basket

Description

Description

Specifications

Specifications

S. aureus Enterotoxin type E Protein
1 mg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
entE
Recombinant Protein
E. coli
SEEINEKDLR KKSELQRNAL SNLRQIYYYN EKAITENKES DDQFLENTLL FKGFFTGHPW YNDLLVDLGS KDATNKYKGK KVDLYGAYYG YQCAGGTPNK TACMYGGVTL HDNNRLTEEK KVPINLWIDG KQTTVPIDKV KTSKKEVTVQ ELDLQARHYL HGKFGLYNSD SFGGKVQRGL IVFHSSEGST VSYDLFDAQG QYPDTLLRIY RDNKTINSEN LHIDLYLYTT
Videos
SDS
Documents
Certificates
Promotions

Promotions