missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Enterotoxin type C-2 Protein

A DNA sequence encoding the Staphylococcus aureus Enterotoxin type C-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7197-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16097650

  • 3405.00 € / 1mg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_001043552.1
1 mg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
S. aureus Enterotoxin type C-2 Protein
Research Use Only
entC2
Recombinant Protein
E. coli
ESQPDPTPDE LHKSSEFTGT MGNMKYLYDD HYVSATKVMS VDKFLAHDLI YNISDKKLKN YDKVKTELLN EDLAKKYKDE VVDVYGSNYY VNCYFSSKDN VGKVTGGKTC MYGGITKHEG NHFDNGNLQN VLIRVYENKR NTISFEVQTD KKSVTAQELD IKARNFLINK KNLYEFNSSP YETGYIKFIE NNGNTFWYDM MPAPGDKFDQ SKYLMMYNDN KTVDSKSVKI EVHLTTKNG
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions