missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Enolase Protein

A DNA sequence encoding the Staphylococcus aureus (strain Mu50 / ATCC 700699) Enolase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7255-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16097870

Quantity Price
1 810.66 € / 100µg
5 762.51 € / 100µg
Estimated Shipment
09-06-2021
Add to basket


Description

Description

Specifications

Specifications

WP_001121760.1
S. aureus Enolase Protein
100 μg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
31213594
Greater than 90% as determined by SDS-PAGE.
Research Use Only
eno
Recombinant Protein
E. coli
MPIITDVYAR EVLDSRGNPT VEVEVLTESG AFGRALVPSG ASTGEHEAVE LRDGDKSRYL GKGVTKAVEN VNEIIAPEII EGEFSVLDQV SIDKMMIALD GTPNKGKLGA NAILGVSIAV ARAAADLLGQ PLYKYLGGFN GKQLPVPMMN IVNGGSHSDA PIAFQEFMIL PVGATTFKES LRWGTEIFHN LKSILSKRGL ETAVGDEGGF APKFEGTEDA VETIIQAIEA AGYKPGEEVF LGFDCASSEF YENGVYDYSK FEGEHGAKRT AAEQVDYLEQ LVDKYPIITI EDGMDENDWD GWKQLTERIG DRVQLVGDDL FVTNTEILAK GIENGIGNSI LIKVNQIGTL TETFDAIEMA QKAGYTAVVS HRSGETEDTT IADIAVATNA GQIKTGSLSR TDRIAKYNQL LRIEDELFET AKYDGIKSFY NLDK
Videos
SDS
Documents