missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Type II iodothyronine deiodinase Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Type II iodothyronine deiodinase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9482-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16098401

  • 770.00 € / 100µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_113908.3
Type II iodothyronine deiodinase Protein
Research Use Only
Dio2
Recombinant Protein
Yeast
MGLLSVDLLI TLQILPVFFS NCLFLALYDS VILLKHVALL LSRSKSTRGE WRRMLTSEGL RCVWNSFLLD AYKQVKLGED APNSSVVHVS NPEAGNNCAS EKTADGAECH LLDFASAERP LVVNFGSATU PPFTRQLPAF RQLVEEFSSV ADFLLVYIDE AHPSDGWAVP GDSSMSFEVK KHRNQEDRCA AAHQLLERFS LPPQCQVVAD RMDNNANVAY GVAFERVCIV QRRKIAYLGG KGPFSYNLQE VRSWLEKNFS KRUILD
Greater than 90% as determined by SDS-PAGE.
65162
100 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions