missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Thyroxine 5-deiodinase Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Thyroxine 5-deiodinase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7221-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16077770

  • 770.00 € / 100µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_058906.3
Thyroxine 5-deiodinase Protein
Research Use Only
Dio3
Recombinant Protein
E. coli
DFLCIRKHFL RRRHPDHPEP EVELNSEGEE MPPDDPPICV SDDNRLCTLA SLKAVWHGQK LDFFKQAHEG GPAPNSEVVR PDGFQSQRIL DYAQGTRPLV LNFGSCTUPP FMARMSAFQR LVTKYQRDVD FLIIYIEEAH PSDGWVTTDS PYVIPQHRSL EDRVSAARVL QQGAPGCALV LDTMANSSSS AYGAYFERLY VIQSGTIMYQ GGRGPDGYQV SELRTWLERY DEQLHGTRPR RL
Greater than 90% as determined by SDS-PAGE.
29475
100 μg
Not determined.
Extracellular Domain
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers