missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Suppressor of cytokine signaling 3 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Suppressor of cytokine signaling 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6713-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16095300

  • 770.00 € / 100µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_446017.1
Suppressor of cytokine signaling 3 Protein
Research Use Only
Socs3
Recombinant Protein
E. coli
MVTHSKFPAA GMSRPLDTSL RLKTFSSKSE YQLVVNAVRK LQESGFYWSA VTGGEANLLL SAEPAGTFLI RDSSDQRHFF TLSVETQSGT KNLRIQCEGG SFSLQSDPRS TQPVPRFDCV LKLVHHYMPP PGAPSFSLPP TEPSFEVQEQ PPAQALPGGT PKRAYYIYSG GEKIPLVLSR PLSSNVATLQ HLCRKTVNGH LDSYEKVTQL PGPIREFLDQ YDAPL
Greater than 90% as determined by SDS-PAGE.
89829
100 μg
Not determined.
Full Length
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions