missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Prolactin-inducible protein homolog Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Prolactin-inducible protein homolog, was expressed in the hosts and tags indicated.
245.00 € - 2645.00 €
Specifications
Accession Number | NP_073199.1 |
---|---|
Gene ID (Entrez) | 64673 |
Name | Prolactin-inducible protein homolog Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048621
|
enQuireBio™
QP9736-YE-100UG |
100 μg |
770.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088621
|
enQuireBio™
QP9736-YE-500UG |
500 μg |
2195.00 €
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058621
|
enQuireBio™
QP9736-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078621
|
enQuireBio™
QP9736-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098621
|
enQuireBio™
QP9736-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068621
|
enQuireBio™
QP9736-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_073199.1 | |
Prolactin-inducible protein homolog Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
64673 | |
Research Use Only | |
Pip | |
Recombinant Protein | |
Yeast | |
QDNETIPQPL LFQLNVPSTP DENQEVDMSL TLQTQYKECL VVKAYLISNT PVDGGFNYIQ TRCICNDHPT TLYWTFVVTQ TLTFRIMVDI VKDKGICPNN VAVVPISGNR YFTDRTVYVN | |
Greater than 90% as determined by SDS-PAGE. |