missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat PLBD2 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) PLBD2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7660-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16070041

  • 3405.00 € / 1mg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_640348.2
PLBD2 Protein
Research Use Only
Plbd2
Recombinant Protein
E. coli
GALPTLGPGW RRQNPEPPAS RTRSLLLDAA SGQLRLEYGF HPDAVAWANL TNAIRETGWA YLDLGTNGSY NDSLQAYAAG VVEASVSEEL IYMHWMNTVV NYCGPFEYEV GYCEKLKSFL EANLEWMQRE MELSPDSPYW HQVRLTLLQL KGLEDSYEGR LTFPTGRFNI KPLGFLLLQI SGDLEDLEPA LNKTNTKPSV GSGSCSALIK LLPGSHDLLV AHNTWNSYQN MLRIIKKYRL QFREGPQEEY PLIAGNNLIF SSYPGTIFSG DDFYILGSGL VTLETTIGNK NPALWKYVQP QGCVLEWIRN IVANRLALDG ATWADVFRRF NSGTYNNQWM IVDYKAFIPN GPSPGSRVLT ILEQIPGMVV VADKTAELYK TTYWASYNIP YFESVFNASG LQALVAQYGD WFSYTRNPRA KIFQRDQSLV EDVDTMVRLM RYNDFLHDPL SLCEACSPKP NAENAISARS DLNPANGSYP FQALRQRAHG GIDVKVTSVA LAKYMSMLAA SGPTWDQLPP FQWSKSPFHN MLHMGQPDLW MFSPVKVPWD
Greater than 90% as determined by SDS-PAGE.
246120
1 mg
Not determined.
Full Length
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions