missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PLAUR / CD87 / uPAR Protein
A DNA sequence encoding the Rattus norvegicus (Rat) PLAUR / CD87 / uPAR, was expressed in the hosts and tags indicated.
245.00 € - 3405.00 €
Specifications
Accession Number | NP_059046.1 |
---|---|
Gene ID (Entrez) | 50692 |
Name | PLAUR / CD87 / uPAR Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16063770
|
enQuireBio™
QP6504-EC-100UG |
100 μg |
770.00 €
100µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16083770
|
enQuireBio™
QP6504-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16073770
|
enQuireBio™
QP6504-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093770
|
enQuireBio™
QP6504-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003780
|
enQuireBio™
QP6504-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013780
|
enQuireBio™
QP6504-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_059046.1 | |
PLAUR / CD87 / uPAR Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
50692 | |
Research Use Only | |
PLAUR | |
Recombinant Protein | |
E. coli | |
LRCIQCESNQ DCLVEECALG QDLCRTTVLR EWEDAEELEV VTRGCAHKEK TNRTMSYRMG SVIVSLTETV CATNLCNRPR PGARGRPFPR GRYLECASCT SLDQSCERGR EQSLQCRYPT EHCIEVVTLQ STERSVKDEP YTKGCGSLPG CPGTAGFHSN QTFHFLKCCN FTQCNGGP | |
Greater than 90% as determined by SDS-PAGE. |