missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat NADPH oxidase 4 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) NADPH oxidase 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9309-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16098071

  • 770.00 € / 100µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_445976.1
NADPH oxidase 4 Protein
Research Use Only
Nox4
Recombinant Protein
Yeast
GGLLKYQTNL DTHPPGCISL NRTPSQNMSI ADYVSEHFHG SLPGGFSKLE DHYQKTLVKI CLEEPKFQAH FPQTWIWISG PLCLYCAERL YRCIRSNKPV TIISVINHPS DVMELRMIKE NFKARPGQYI ILHCPSVSAL ENHPFTLTMC PTETKATFGV HFKVVGDWTE RFRDLLLPPS SQDSEILPFI QSRNYPKLYI DGPFGSPFEE SLNYE
Greater than 90% as determined by SDS-PAGE.
85431
100 μg
Not determined.
Extracellular Domain
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions