missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Cytochrome P450 1B1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Cytochrome P450 1B1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7721-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16070411

Quantity Price
1 725.49 € / 100µg
5 682.62 € / 100µg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_037072.1
Cytochrome P450 1B1 Protein
100 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
25426
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Cyp1b1
Recombinant Protein
E. coli
MATSLSADSP QQLSSLSTQQ TILLLLVSVL AIVHLGQWLL RQWRRKPWSS PPGPFPWPLI GNAASVGRAS HLYFARLARR YGDVFQIRLG SCPVVVLNGE SAIHQALVQQ GGVFADRPPF ASFRVVSGGR SLAFGHYSER WKERRRAAYG TMRAFSTRHP RSRGLLEGHA LGEARELVAV LVRRCAGGAC LDPTQPIIVA VANVMSAVCF GCRYNHDDAE FLELLSHNEE FGRTVGAGSL VDVMPWLQLF PNPVRTIFRE FEQINRNFSN FVLDKFLRHR ESLVPGAAPR DMMDAFILSA EKKATGDPGD SPSGLDLEDV PATITDIFGA SQDTLSTALL WLLILFTRYP DVQARVQAEL DQVVGRDRLP CMSDQPNLPY VMAFLYESMR FTSFLPVTLP HATTANTFVL GYYIPKNTVV FVNQWSVNHD PAKWSNPEDF DPARFLDKDG FINKALASSV MIFSVGKRRC IGEELSKTLL FLFISILAHQ CNFKANQNEP SNMSFSYGLS IKPKSFKIHV SLRESMKLLD SAVEKLQAEE ACQ
Videos
SDS
Documents
Certificates
Promotions

Promotions