missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat BDNF Protein Isoform 2 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) BDNF Protein Isoform 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8869-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16076911

  • 3405.00 € / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001257559.1
BDNF Protein Isoform 2 Protein
Research Use Only
BDNF
Recombinant Protein
E. coli
RRGELSVCDS ISEWVTAADK KTAVDMSGGT VTVLEKVPVS KGQLKQYFYE TKCNPMGYTK EGCRGIDKRH WNSQCRTTQS YVRALTMDSK KRIGWRFIRI DTSCVCTL
Greater than 90% as determined by SDS-PAGE.
24225
1 mg
Not determined.
Partial (see sequence information for more details).
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions