missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant P. gingivalis Gingipain R1 Protein

A DNA sequence encoding the Porphyromonas gingivalis Gingipain R1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7192-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16087630

  • 3405.00 € / 1mg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

P. gingivalis Gingipain R1 Protein
Research Use Only
rgpA
Recombinant Protein
E. coli
YTPVEEKQNG RMIVIVAKKY EGDIKDFVDW KNQRGLRTEV KVAEDIASPV TANAIQQFVK QEYEKEGNDL TYVLLVGDHK DIPAKITPGI KSDQVYGQIV GNDHYNEVFI GRFSCESKED LKTQIDRTIH YERNITTEDK WLGQALCIAS AEGGPSADNG ESDIQHENVI ANLLTQYGYT KIIKCYDPGV TPKNIIDAFN GGISLVNYTG HGSETAWGTS HFGTTHVKQL TNSNQLPFIF DVACVNGDFL FSMPCFAEAL MRAQKDGKPT GTVAIIASTI NQSWASPMRG QDEMNEILCE KHPNNIKRTF GGVTMNGMFA MVEKYKKDGE KMLDTWTVFG DPSLLVRTLV PTKMQVTAPA QINLTDASVN VSCDYNGAIA TISANGKMFG SAVVENGTAT INLTGLTNES TLTLTVVGYN KETVIKTINT NGEPNPYQPV SNLTATTQGQ KVTLKWDAPS TKTNATTNTA RSVDGIRELV LLSVSDAPEL LRS
Greater than 80% as determined by SDS-PAGE.
1 mg
Not determined.
Partial (see sequence information for more details).
P. gingivalis
His-B2M
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions