missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Wdpcp Protein
A DNA sequence encoding the Mus musculus (Mouse) Wdpcp, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7804-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_663400.2 | |
Wdpcp Protein | |
Research Use Only | |
Wdpcp | |
Recombinant Protein | |
E. coli | |
MSFCLTELHL WSLKSTLHIA DRDIGVYQYY DKKDPSVSAT EHGNLEEKQR LAESRDYPWT LKNRRPEKLR DSLKELEELM QSSPCVLSKW KSKYICQLLF GSGVLVSLSL SGPQLEKVVI DRSLVGKLIS DTISDALLTD SFIILSFLAQ NKLCFIQFTK KMDSLDGNKR LEKLSALDLK ISYYDIPGPA NRTIDRHLAV NSTQDLVVCW WPLVSDDVWP WTPVSSEKDR ANMLLLGFTQ GGLEVLSFVR TEWSPLDVHF GTKQPYQVFT VECSVSVDKE PMADSCIYES VRNKLHCVSV TRIPLRSKAI SCCRNSTEDK LIVGCEDSSV ILYEAHRGVT LLAQAELRPS LISCHPSGAI LLVGSNQGEL QIFDIALSPI NIQLLAEDYS PKETLQFKKF FDVSSSLVQM QWMAPPVVFQ KPKRGEICDL LFLRFNKGPL GVLLFKLGIL TRGQLGLVDL ILQYIHYSEV YEAISILRSM DWDTLGQQCL IGMGTIVNHL LRQRLTPERE AQLEASLGTF YAPTRPLLDT TILEYREPVS KYARRLFHHL LRYKRFEKAF LLAVDIGARD LFMDIHYLAL DMGELALAEV ARRRAHDIDV ESVSSGVELL GPLDRRDMLN EGFASSALMP EGENKFPGLL PSIGSTHMQT LQQKIPNGPS SRWAIERRTE EEEEEEEEEE EELCTDSSGA TTWNAEGELK EDQRKQDIGD VGSLRMVHFG LV | |
Greater than 90% as determined by SDS-PAGE. |
216560 | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |