missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Wdpcp Protein
A DNA sequence encoding the Mus musculus (Mouse) Wdpcp, was expressed in the hosts and tags indicated.
245.00 € - 3405.00 €
Specifications
Accession Number | NP_663400.2 |
---|---|
Gene ID (Entrez) | 216560 |
Name | Wdpcp Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16090891
|
enQuireBio™
QP7804-EC-100UG |
100 μg |
770.00 €
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16010901
|
enQuireBio™
QP7804-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16000901
|
enQuireBio™
QP7804-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16020901
|
enQuireBio™
QP7804-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030901
|
enQuireBio™
QP7804-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16040901
|
enQuireBio™
QP7804-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_663400.2 | |
Wdpcp Protein | |
Not determined. | |
Full Length | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |
216560 | |
Research Use Only | |
Wdpcp | |
Recombinant Protein | |
E. coli | |
MSFCLTELHL WSLKSTLHIA DRDIGVYQYY DKKDPSVSAT EHGNLEEKQR LAESRDYPWT LKNRRPEKLR DSLKELEELM QSSPCVLSKW KSKYICQLLF GSGVLVSLSL SGPQLEKVVI DRSLVGKLIS DTISDALLTD SFIILSFLAQ NKLCFIQFTK KMDSLDGNKR LEKLSALDLK ISYYDIPGPA NRTIDRHLAV NSTQDLVVCW WPLVSDDVWP WTPVSSEKDR ANMLLLGFTQ GGLEVLSFVR TEWSPLDVHF GTKQPYQVFT VECSVSVDKE PMADSCIYES VRNKLHCVSV TRIPLRSKAI SCCRNSTEDK LIVGCEDSSV ILYEAHRGVT LLAQAELRPS LISCHPSGAI LLVGSNQGEL QIFDIALSPI NIQLLAEDYS PKETLQFKKF FDVSSSLVQM QWMAPPVVFQ KPKRGEICDL LFLRFNKGPL GVLLFKLGIL TRGQLGLVDL ILQYIHYSEV YEAISILRSM DWDTLGQQCL IGMGTIVNHL LRQRLTPERE AQLEASLGTF YAPTRPLLDT TILEYREPVS KYARRLFHHL LRYKRFEKAF LLAVDIGARD LFMDIHYLAL DMGELALAEV ARRRAHDIDV ESVSSGVELL GPLDRRDMLN EGFASSALMP EGENKFPGLL PSIGSTHMQT LQQKIPNGPS SRWAIERRTE EEEEEEEEEE EELCTDSSGA TTWNAEGELK EDQRKQDIGD VGSLRMVHFG LV | |
Greater than 90% as determined by SDS-PAGE. |