missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse VEGF-C Protein
A DNA sequence encoding the Mus musculus (Mouse) VEGF-C, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6880-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_033532.1 | |
VEGF-C Protein | |
Research Use Only | |
Vegfc | |
Recombinant Protein | |
E. coli | |
AHYNTEILKS IDNEWRKTQC MPREVCIDVG KEFGAATNTF FKPPCVSVYR CGGCCNSEGL QCMNTSTGYL SKTLFEITVP LSQGPKPVTI SFANHTSCRC MSKLDVYRQV HSIIRR | |
Greater than 90% as determined by SDS-PAGE. |
22341 | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |