missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Tyrosine-protein kinase JAK1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Tyrosine-protein kinase JAK1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6245-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051890

  • 3375.00 € / 1mg

Estimated Shipment: 18-11-2022
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Tyrosine-protein kinase JAK1 Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
JAK1
Recombinant Protein
E. coli
EEQNPDIVSE KQPTTEVDPT HFEKRFLKRI RDLGEGHFGK VELCRYDPEG DNTGEQVAVK SLKPESGGNH IADLKKEIEI LRNLYHENIV KYKGICMEDG GNGIKLIMEF LPSGSLKEYL PKNKNKINLK QQLKYAIQIC KGMDYLGSRQ YVHRDLAARN VLVESEHQVK IGDFGLTKAI ETDKEYYTVK DDRDSPVFWY APECLIQCKF YIASDVWSFG VTLHELLTYC DSDFSPMALF LKMIGPTHGQ MTVTRLVKTL KEGKRLPCPP NCPDEVYQLM RKCWEFQPSN RTTFQNLIEG FEALL
Videos
SDS
Documents
Certificates
Promotions

Promotions