missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Troponin I, cardiac muscle Protein

A DNA sequence encoding the Mus musculus (Mouse) Troponin I, cardiac muscle, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7213-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16097740

  • 3405.00 € / 1mg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_033432.1
Troponin I, cardiac muscle Protein
Research Use Only
Tnni3
Recombinant Protein
E. coli
ADESSDAAGE PQPAPAPVRR RSSANYRAYA TEPHAKKKSK ISASRKLQLK TLMLQIAKQE MEREAEERRG EKGRVLRTRC QPLELDGLGF EELQDLCRQL HARVDKVDEE RYDVEAKVTK NITEIADLTQ KIYDLRGKFK RPTLRRVRIS ADAMMQALLG TRAKESLDLR AHLKQVKKED IEKENREVGD WRKNIDALSG MEGRKKKFEG
Greater than 82.9% as determined by SDS-PAGE.
21954
1 mg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions