missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Ripk3 Protein

A DNA sequence encoding the Mus musculus (Mouse) Ripk3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9919-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16089001

Quantity Price
1 725.49 € / 100µg
5 682.62 € / 100µg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_064339.2
Ripk3 Protein
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
56532
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Ripk3
Recombinant Protein
Yeast
MSSVKLWPTG ASAVPLVSRE ELKKLEFVGK GGFGVVFRAH HRTWNHDVAV KIVNSKKISW EVKAMVNLRN ENVLLLLGVT EDLQWDFVSG QALVTRFMEN GSLAGLLQPE CPRPWPLLCR LLQEVVLGMC YLHSLDPPLL HRDLKPSNIL LDPELHAKLA DFGLSTFQGG SQSGSGSGSG SRDSGGTLAY LDPELLFKVN LKASKASDVY SFGILVWAVL AGREAELVDK TSLIRETVCD RQSRPPLTEL PPGSPETPGL EKLKELMIHC WGSQSENRPS FQDCEPKTNE VYNLVKDKVD AAVSEVKHYL SQHRSSGRNL SAREPSQRGT EMDCPRETMV SKMLDRLHLE EPSGPVPGKC PERQAQDTSV GPATPARTSS DPVAGTPQIP HTLPFRGTTP GPVFTETPGP HPQRNQGDGR HGTPWYPWTP PNPMTGPPAL VFNNCSEVQI GNYNSLVAPP RTTASSSAKY DQAQFGRGRG WQPFHK
Videos
SDS
Documents
Certificates
Promotions

Promotions