missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Protein ABHD11 Protein

A DNA sequence encoding the Mus musculus (Mouse) Protein ABHD11, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7815-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16090951

  • 725.49 € / 100µg

Estimated Shipment: 10-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_660250.1
Protein ABHD11 Protein
100 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
68758
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Abhd11
Recombinant Protein
E. coli
MLRWARAWRV PRGVLGASSP RRLAVPVTFC SSRSSGQENA DLRPLPLSYN LLDGDATLPA IVFLHGLFGS KTNFNSLAKA MVQRTGRRVL TVDARNHGDS PHSPDASYEA MSQDLQGLLP QLGLVPCVLV GHSMGGKTAM LLALQRPDVV ERLVVVDISP VGTTPGSHIG AFIAAMKAVE IPEKVPHSQA RKLADKQLSS VVKEAGIRQF LLTNLVEVGG RFSWRLNLDT LAQHLDKIMT FPQQREPYSG PTLFLLGGNS TYVQPSHHSE IRRLFPQAQI QTVPNAGHWV HSDKPQDFMD AVTSFLA
Videos
SDS
Documents
Certificates
Promotions

Promotions