missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse MSLN / Mesothelin Protein

A DNA sequence encoding the Mus musculus (Mouse) MSLN / Mesothelin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6389-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16082950

  • 725.49 € / 100µg

Estimated Shipment: 10-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_061345.1
MSLN / Mesothelin Protein
100 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
56047
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Msln
Recombinant Protein
E. coli
DAEQKACPPG KEPYKVDEDL IFYQNWELEA CVDGTMLARQ MDLVNEIPFT YEQLSIFKHK LDKTYPQGYP ESLIQQLGHF FRYVSPEDIH QWNVTSPDTV KTLLKVSKGQ KMNAQAIALV ACYLRGGGQL DEDMVKALGD IPLSYLCDFS PQDLHSVPSS VMWLVGPQDL DKCSQRHLGL LYQKACSAFQ NVSGLEYFEK IKTFLGGASV KDLRALSQHN VSMDIATFKR LQVDSLVGLS VAEVQKLLGP NIVDLKTEED KSPVRDWLFR QHQKDLDRLG LGLQGGIPNG YLVLDFNVRE AFS
Videos
SDS
Documents
Certificates
Promotions

Promotions