missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Microtubule-associated protein 1
A DNA sequence encoding the Mus musculus (Mouse) Microtubule-associated protein 2, was expressed in the hosts and tags indicated.
245.00 € - 3405.00 €
Specifications
Accession Number | NP_001297563.1 |
---|---|
Gene ID (Entrez) | 17756 |
Name | Microtubule-associated protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16092550
|
enQuireBio™
QP6335-EC-100UG |
100 μg |
770.00 €
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16012560
|
enQuireBio™
QP6335-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16002560
|
enQuireBio™
QP6335-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16022560
|
enQuireBio™
QP6335-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16032560
|
enQuireBio™
QP6335-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16042560
|
enQuireBio™
QP6335-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001297563.1 | |
Microtubule-associated protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
17756 | |
Research Use Only | |
Map2 | |
Recombinant Protein | |
E. coli | |
GMEGEKLPPV PFAQTFGTNL EDRKQSTEPS IVMPSIGLSA EPPAPKEPKD WFIEMPTESK KDEWGLAAPI SPGPLTPMRE KDVLEDIPRW EGKQFDSPMP SPFHGGSFTL PLDTMKNERV SEGPRPFAPV FFQSDDKVSL QDPSALATSK ESSKDEEPLK DKADKVADVS ISEVTTLLGN VHSPVVEGYV GENISGEVKV TTDQEKKETS APSVQEPTLT ETEPQTKLDE KSTVSIEEAV AKKEESFKLR DDKTGVIQTS TEQSFSKEDQ KGQEHTIDEL KQDSFPISLE QAVTDAAMTS KTLGKVTSEP EAVSERREIQ GLFEEKTADK NKLEGAGSAT IAEVEMP | |
Greater than 90% as determined by SDS-PAGE. |