missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Lingo1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Lingo1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8168-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16093021

  • 770.00 € / 100µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001298005.1
Lingo1 Protein
Research Use Only
Lingo1
Recombinant Protein
Yeast
PPRCECSAQD RAVLCHRKRF VAVPEGIPTE TRLLDLGKNR IKTLNQDEFA SFPHLEELEL NENIVSAVEP GAFNNLFNLR TLGLRSNRLK LIPLGVFTGL SNLTKLDISE NKIVILLDYM FQDLYNLKSL EVGDNDLVYI SHRAFSGLNS LEQLTLEKCN LTSIPTEALS HLHGLIVLRL RHLNINAIRD YSFKRLYRLK VLEISHWPYL DTMTPNCLYG LNLTSLSITH CNLTAVPYLA VRHLVYLRFL NLSYNPIGTI EGSMLHELLR LQEIQLVGGQ LAVVEPYAFR GLNYLRVLNV SGNQLTTLEE SAFHSVGNLE TLILDSNPLA CDCRLLWVFR RRWRLNFNRQ QPTCATPEFV QGKEFKDFPD VLLPNYFTCR RAHIRDRKAQ QVFVDEGHTV QFVCRADGDP PPAILWLSPR KHLVSAKSNG RLTVFPDGTL EVRYAQVQDN GTYLCIAANA GGNDSMPAHL HVRSYSPDWP HQPNKTFAFI SNQPGEGEAN STRATVPFPF DIKTLIIAT
Greater than 80% as determined by SDS-PAGE.
235402
100 μg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions