missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Fibroblast growth factor 15 Protein

A DNA sequence encoding the Mus musculus (Mouse) Fibroblast growth factor 15, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7474-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16088930

  • 770.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_032029.1
Fibroblast growth factor 15 Protein
Research Use Only
Fgf15
Recombinant Protein
E. coli
RPLAQQSQSV SDEDPLFLYG WGKITRLQYL YSAGPYVSNC FLRIRSDGSV DCEEDQNERN LLEFRAVALK TIAIKDVSSV RYLCMSADGK IYGLIRYSEE DCTFREEMDC LGYNQYRSMK HHLHIIFIQA KPREQLQDQK PSNFIPVFHR SFFETGDQLR SKMFSLPLES DSMDPFRMVE DVDHLVKSPS FQK
Greater than 90% as determined by SDS-PAGE.
14170
100 μg
Not determined.
Full Length of Mature Protein
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions