missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CMPK1 Protein
A DNA sequence encoding the Mus musculus (Mouse) CMPK1, was expressed in the hosts and tags indicated.
309.00 € - 3405.00 €
Specifications
Accession Number | NP_079923.3 |
---|---|
Gene ID (Entrez) | 66588 |
Name | CMPK1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16092841
|
enQuireBio™
QP8136-EC-100UG |
100 μg |
985.00 €
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16012851
|
enQuireBio™
QP8136-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16002851
|
enQuireBio™
QP8136-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16022851
|
enQuireBio™
QP8136-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16032851
|
enQuireBio™
QP8136-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16042851
|
enQuireBio™
QP8136-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_079923.3 | |
CMPK1 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
66588 | |
Research Use Only | |
Cmpk1 | |
Recombinant Protein | |
E. coli | |
MKPLVVFVLG GPGAGKGTQC ARIVEKYGYT HLSAGELLRD ERKNPDSQYG ELIEKYIKEG KIVPVEITIS LLKREMDQTM AANAQKNKFL IDGFPRNQDN LQGWNKTMDG KADVSFVLFF DCNNEICIER CLERGKSSGR SDDNRESLEK RIQTYLESTK PIIDLYEEMG KVKKIDASKS VDEVFGEVVK IFDKEG | |
Greater than 90% as determined by SDS-PAGE. |