missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Arginase 1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Arginase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7724-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16090421

  • 725.49 € / 100µg

Estimated Shipment: 10-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_031508.1
Arginase 1 Protein
100 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
11846
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ARG1
Recombinant Protein
E. coli
MSSKPKSLEI IGAPFSKGQP RGGVEKGPAA LRKAGLLEKL KETEYDVRDH GDLAFVDVPN DSSFQIVKNP RSVGKANEEL AGVVAEVQKN GRVSVVLGGD HSLAVGSISG HARVHPDLCV IWVDAHTDIN TPLTTSSGNL HGQPVSFLLK ELKGKFPDVP GFSWVTPCIS AKDIVYIGLR DVDPGEHYII KTLGIKYFSM TEVDKLGIGK VMEETFSYLL GRKKRPIHLS FDVDGLDPAF TPATGTPVLG GLSYREGLYI TEEIYKTGLL SGLDIMEVNP TLGKTAEEVK STVNTAVALT LACFGTQREG NHKPGTDYLK PPK
Videos
SDS
Documents
Certificates
Promotions

Promotions