missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant MERS-CoV Spike RBD His-tag Protein, CF
HEK293 Expressed
Brand: R&D Systems™ 10636-CV-100
This item is not returnable.
View return policy
Description
Measured by its binding ability in a functional ELISA with Recombinant Human DPPIV/CD26 (High Purity Dimer) (Catalog # 9168-SE ).Specifications
YP_007188579.1 | |
Supplied as a 0.2 μm filtered solution in PBS with Trehalose. | |
100 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after opening. 3 months, -20 to -70°C under sterile conditions after opening. | |
Unconjugated | |
Spike RBD |
Bioactivity | |
MolecularWeight-observed: 34-41 kDa, under reducing conditions, MolecularWeight-theroretical: 27 kDa | |
Human embryonic kidney cell, HEK293-derived mers-cov Spike RBD protein Glu367-Tyr606, with a C-terminal 6-His tag | |
<0.10 EU per 1 μg of the protein by the LAL method. | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction