missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant M. tuberculosis Steroid C26-monooxygenase Protein
A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Steroid C26-monooxygenase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7253-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_218062.1 | |
M. tuberculosis Steroid C26-monooxygenase Protein | |
Research Use Only | |
cyp125 | |
Recombinant Protein | |
Yeast | |
MSWNHQSVEI AVRRTTVPSP NLPPGFDFTD PAIYAERLPV AEFAELRSAA PIWWNGQDPG KGGGFHDGGF WAITKLNDVK EISRHSDVFS SYENGVIPRF KNDIAREDIE VQRFVMLNMD APHHTRLRKI ISRGFTPRAV GRLHDELQER AQKIAAEAAA AGSGDFVEQV SCELPLQAIA GLLGVPQEDR GKLFHWSNEM TGNEDPEYAH IDPKASSAEL IGYAMKMAEE KAKNPADDIV TQLIQADIDG EKLSDDEFGF FVVMLAVAGN ETTRNSITQG MMAFAEHPDQ WELYKKVRPE TAADEIVRWA TPVTAFQRTA LRDYELSGVQ IKKGQRVVMF YRSANFDEEV FQDPFTFNIL RNPNPHVGFG GTGAHYCIGA NLARMTINLI FNAVADHMPD LKPISAPERL RSGWLNGIKH WQVDYTGRCP VAH | |
Greater than 90% as determined by SDS-PAGE. |
887782 | |
500 μg | |
Not determined. | |
Full Length | |
M. tuberculosis | |
His | |
Tris-based buffer, 50% glycerol |