missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant M. tuberculosis Ag85C Protein

A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Ag85C, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7382-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16068560

Quantity Price
1 920.10 € / 100µg
5 865.45 € / 100µg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

WP_003400908.1
Greater than 90% as determined by SDS-PAGE.
Research Use Only
fbpC
Recombinant Protein
E. coli
AFSRPGLPVE YLQVPSASMG RDIKVQFQGG GPHAVYLLDG LRAQDDYNGW DINTPAFEEY YQSGLSVIMP VGGQSSFYTD WYQPSQSNGQ NYTYKWETFL TREMPAWLQA NKGVSPTGNA AVGLSMSGGS ALILAAYYPQ QFPYAASLSG FLNPSEGWWP TLIGLAMNDS GGYNANSMWG PSSDPAWKRN DPMVQIPRLV ANNTRIWVYC GNGTPSDLGG DNIPAKFLEG LTLRTNQTFR DTYAADGGRN GVFNFPPNGT HSWPYWNEQL VAMKADIQHV LNGATPPAAP AAPAA
M. tuberculosis Ag85C Protein
100 μg
Not determined.
Full Length
M. tuberculosis
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents
Certificates
Promotions

Promotions