missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H3N2 (Kitakyushu / 159 / 1993) NucleoProtein
A DNA sequence encoding the Influenza A virus (strain A/Kitakyushu/159/1993 H3N2) Influenza A H3N2 (strain A/Kitakyushu/159/1993) Nucleoprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7500-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
Influenza A H3N2 (Kitakyushu / 159 / 1993) NucleoProtein | |
Research Use Only | |
NP | |
Recombinant Protein | |
E. coli | |
MASQGTKRSY EQMETDGERQ NATEIRASVG KMIDGIGRFY IQMCTELKLS DYEGRLIQNS LTIERMVLSA FDERRNRYLE EHPSAGKDPK KTGGPIYKRV DGRWMRELVL YDKEEIRRIW RQANNGDDAT AGLTHMMIWH SNLNDTTYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGIGTMV MELIRMIKRG INDRNFWRGE NGRKTRSAYE RMCNILKGKF QTAAQRAMMD QVRESRNPGN AEIEDLIFSA RSALILRGSV AHKSCLPACV YGPAVSSGYN FEKEGYSLVG IDPFKLLQNS QVYSLIRPNE NPAHKSQLVW MACHSAAFED LRLLSFIRGT KVSPRGKLST RGVQIASNEN MDNMESSTLE LRSRYWAIRT RSGGNTNQQR ASAGQISVQP TFSVQRNLPF EKSTVMAAFT GNTEGRTSDM RAEIIRMMEG AKPEEVSFRG RGVFELSDEK ATNPIVPSFD MSNEGSYFFG DNAEEYDN | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
Influenza | |
His | |
Tris-based buffer, 50% glycerol |