missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza A H3N2 (Hong Kong / 1 / 1968) Nucleoprotein / NP Protein

A DNA sequence encoding the Influenza A virus (strain A/Hong Kong/1/1968 H3N2) Influenza A H3N2 (A/Hong Kong/1/1968) Nucleoprotein / NP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7093-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16087360

  • 440.00 € / 10µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Influenza A H3N2 (Hong Kong / 1 / 1968) Nucleoprotein / NP Protein
Research Use Only
NP
Recombinant Protein
E. coli
MASQGTKRSY EQMETDGERQ NATEIRASVG KMIDGIGRFY IQMCTELKLS DYEGRLIQNS LTIERMVLSA FDERRNKYLE EHPSAGKDPK KTGGPIYKRV DGKWMRELVL YDKEEIRRIW RQANNGDDAT AGLTHMMIWH SNLNDTTYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGVGTMV MELIRMIKRG INDRNFWRGE NGRKTRSAYE RMCNILKGKF QTAAQRAMMD QVRESRNPGN AEIEDLIFLA RSALILRGSV AHKSCLPACV YGPAVASGYD FEKEGYSLVG IDPFKLLQNS QVYSLIRPNE NPAHKSQLVW MACNSAAFED LRVLSFIRGT KVSPRGKLST RGVQIASNEN MDAMESSTLE LRSRYWAIRT RSGGNTNQQR ASAGQISVQP AFSVQRNLPF DKPTIMAAFT GNTEGRTSDM RAEIIRMMEG AKPEEMSFQG RGVFELSDEK AANPIVPSFD MSNEGSYFFG DNAEEYDN
Greater than 90% as determined by SDS-PAGE.
10 μg
Not determined.
Full Length
Influenza
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions