missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza A H1N1 (Fort Warren / 1 / 1950) Nucleocapsid Protein

A DNA sequence encoding the Influenza A virus (strain A/Fort Warren/1/1950 H1N1) Influenza A H1N1 (strain A/Fort Warren/1/1950) Nucleocapsid, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7081-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16087320

  • 630.00 € / 50µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Influenza A H1N1 (Fort Warren / 1 / 1950) Nucleocapsid Protein
Research Use Only
NP
Recombinant Protein
E. coli
MASQGTKRSY EQMETDGDRQ NATEIRASVG KMIDGIGRFY IQMCTELKLS DYEGRLIQNS LTIERMVLSA FDERRNKYLE EHPSAGKDPK KTGGPIYKRV DGKWMRELVL YDKEEIRRIW RQANNGDDAT AGLTHMMIWH SNLNDTTYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGVGTMV MELIRMIKRG INDRNFWRGE NGRKTRIAYE RMCNILKGKF QTAAQRAMMD QVRESRNPGN AEIEDLIFLA RSALILRGSV AHKSCLPACV YGPAVASGYD FEKEGYSLVG IDPFKLLQNS QVYSLIRPNE NPAHKSQLVW MACNSAAFED LRVSSFIRGT KVIPRGKLST RGVQIASNEN MDTMGSSTLE LRSRYWAIRT RSGGNTNQQR ASAGQISIQP TFSVQRNLPF DKTTIMAAFT GNAEGRTSDM RAEIIRMMES ARPEEVSFQG RGVFELSDER AANPIVPSFD MSNEGSYFFG DNAEEYDN
Greater than 90% as determined by SDS-PAGE.
50 μg
Not determined.
Full Length
Influenza
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions