missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger matrin-type protein 1
A DNA sequence encoding the Homo sapiens (Human) Zinc finger matrin-type protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8767-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_653324.1 | |
Zinc finger matrin-type protein 1 | |
Research Use Only | |
ZMAT2 | |
Recombinant Protein | |
E. coli | |
MASGSGTKNL DFRRKWDKDE YEKLAEKRLT EEREKKDGKP VQPVKRELLR HRDYKVDLES KLGKTIVITK TTPQSEMGGY YCNVCDCVVK DSINFLDHIN GKKHQRNLGM SMRVERSTLD QVKKRFEVNK KKMEEKQKDY DFEERMKELR EEEEKAKAYK KEKQKEKKRR AEEDLTFEED DEMAAVMGFS GFGS | |
Greater than 90% as determined by SDS-PAGE. |
153527 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |