missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger matrin-type protein 1
A DNA sequence encoding the Homo sapiens (Human) Zinc finger matrin-type protein 2, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_653324.1 |
---|---|
Gene ID (Entrez) | 153527 |
Name | Zinc finger matrin-type protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16086371
|
enQuireBio™
QP8767-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16006381
|
enQuireBio™
QP8767-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16096371
|
enQuireBio™
QP8767-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016381
|
enQuireBio™
QP8767-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026381
|
enQuireBio™
QP8767-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036381
|
enQuireBio™
QP8767-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_653324.1 | |
Zinc finger matrin-type protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
153527 | |
Research Use Only | |
ZMAT2 | |
Recombinant Protein | |
E. coli | |
MASGSGTKNL DFRRKWDKDE YEKLAEKRLT EEREKKDGKP VQPVKRELLR HRDYKVDLES KLGKTIVITK TTPQSEMGGY YCNVCDCVVK DSINFLDHIN GKKHQRNLGM SMRVERSTLD QVKKRFEVNK KKMEEKQKDY DFEERMKELR EEEEKAKAYK KEKQKEKKRR AEEDLTFEED DEMAAVMGFS GFGS | |
Greater than 90% as determined by SDS-PAGE. |