missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VEGFR2 / Flk-1 / CD309 / KDR Protein
A DNA sequence encoding the Homo sapiens (Human) VEGFR2 / Flk-1 / CD309 / KDR, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_002244.1 |
---|---|
Gene ID (Entrez) | 3791 |
Name | VEGFR2 / Flk-1 / CD309 / KDR Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091980
|
enQuireBio™
QP6258-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16011990
|
enQuireBio™
QP6258-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001990
|
enQuireBio™
QP6258-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021990
|
enQuireBio™
QP6258-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031990
|
enQuireBio™
QP6258-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041990
|
enQuireBio™
QP6258-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002244.1 | |
VEGFR2 / Flk-1 / CD309 / KDR Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
3791 | |
Research Use Only | |
KDR | |
Recombinant Protein | |
E. coli | |
ASVGLPSVSL DLPRLSIQKD ILTIKANTTL QITCRGQRDL DWLWPNNQSG SEQRVEVTEC SDGLFCKTLT IPKVIGNDTG AYKCFYRETD LASVIYVYVQ DYRSPFIASV SDQHGVVYIT ENKNKTVVIP CLGSISNLNV SLCARYPEKR FVPDGNRISW DSKKGFTIPS YMISYAGMVF CEAKINDESY QSIMYIVVVV GYRIYDVVLS PSHGIELSVG EKLVLNCTAR TELNVGIDFN WEYPSSKHQH KKLVNRDLKT QSGSEMKKFL STLTIDGVTR SDQGLYTCAA SSGLMTKKNS TFVRVHEKPF VAFGSGMESL VEATVGERVR IPAKYLGYPP PEIKWYKNGI PLESNHTIKA GHVLTIMEVS ERDTGNYTVI LTNPISKEKQ SHVVSLVVYV PPQIGEKSLI SPVDSYQYGT TQTLTCTVYA IPPPHHIHWY WQLEEECANE PSQAVSVTNP YPCEEWRSVE DFQGGNKIEV NKNQFALIEG KNKTVSTLVI QAANVSALYK CEAVNKVGRG ERVISFHVTR GPEITLQPDM QPTEQESVSL WCTADRSTFE NLTWYKLGPQ PLPIHVGELP TPVCKNLDTL WKLNATMFSN STNDILIMEL KNASLQDQGD YVCLAQDRKT KKRHCVVRQL TVLERVAPTI TGNLENQTTS IGESIEVSCT ASGNPPPQIM WFKDNETLVE DSGIVLKDGN RNLTIRRVRK EDEGLYTCQA CSVLGCAKVE AFFIIEGAQE KTNLE | |
Greater than 90% as determined by SDS-PAGE. |