missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human VCL / Vinculin Protein

A DNA sequence encoding the Homo sapiens (Human) VCL / Vinculin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8773-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16086401

Quantity Price
1 493.03 € / 100µg
6 463.44 € / 100µg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_003364.1
VCL / Vinculin Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
7414
Greater than 90% as determined by SDS-PAGE.
Research Use Only
VCL
Recombinant Protein
E. coli
PVFHTRTIES ILEPVAQQIS HLVIMHEEGE VDGKAIPDLT APVAAVQAAV SNLVRVGKET VQTTEDQILK RDMPPAFIKV ENACTKLVQA AQMLQSDPYS VPARDYLIDG SRGILSGTSD LLLTFDEAEV RKIIRVCKGI LEYLTVAEVV ETMEDLVTYT KNLGPGMTKM AKMIDERQQE LTHQEHRVML VNSMNTVKEL LPVLISAMKI FVTTKNSKNQ GIEEALKNRN FTVE
Videos
SDS
Documents
Certificates
Promotions

Promotions