Learn More
enQuireBio™ Recombinant Human UTP--glucose-1-phosphate uridylyltransferase Protein
A DNA sequence encoding the Homo sapiens (Human) UTP--glucose-1-phosphate uridylyltransferase, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7578-ec-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16089520
  • 2385.00 € / 1mg
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Specifications
NP_001001521.1
UTP--glucose-1-phosphate uridylyltransferase Protein
Research Use Only
UGP2
Recombinant Protein
E. coli
MSQDGASQFQ EVIRQELELS VKKELEKILT TASSHEFEHT KKDLDGFRKL FHRFLQEKGP SVDWGKIQRP PEDSIQPYEK IKARGLPDNI SSVLNKLVVV KLNGGLGTSM GCKGPKSLIG VRNENTFLDL TVQQIEHLNK TYNTDVPLVL MNSFNTDEDT KKILQKYNHC RVKIYTFNQS RYPRINKESL LPVAKDVSYS GENTEAWYPP GHGDIYASFY NSGLLDTFIG EGKEYIFVSN IDNLGATVDL YILNHLMNPP NGKRCEFVME VTNKTRADVK GGTLTQYEGK LRLVEIAQVP KAHVDEFKSV SKFKIFNTNN LWISLAAVKR LQEQNAIDME IIVNAKTLDG GLNVIQLETA VGAAIKSFEN SLGINVPRSR FLPVKTTSDL LLVMSNLYSL NAGSLTMSEK REFPTVPLVK LGSSFTKVQD YLRRFESIPD MLELDHLTVS GDVTFGKNVS LKGTVIIIAN HGDRIDIPPG AVLENKIVSG NLRILDH
Greater than 90% as determined by SDS-PAGE.
7360
1 mg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol