Learn More
enQuireBio™ Recombinant Human UTP--glucose-1-phosphate uridylyltransferase Protein
A DNA sequence encoding the Homo sapiens (Human) UTP--glucose-1-phosphate uridylyltransferase, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_001001521.1 |
---|---|
Gene ID (Entrez) | 7360 |
Name | UTP--glucose-1-phosphate uridylyltransferase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16069520
|
enQuireBio™
QP7578-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16089520
|
enQuireBio™
QP7578-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16079520
|
enQuireBio™
QP7578-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099520
|
enQuireBio™
QP7578-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009530
|
enQuireBio™
QP7578-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16019530
|
enQuireBio™
QP7578-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001001521.1 | |
UTP--glucose-1-phosphate uridylyltransferase Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
7360 | |
Research Use Only | |
UGP2 | |
Recombinant Protein | |
E. coli | |
MSQDGASQFQ EVIRQELELS VKKELEKILT TASSHEFEHT KKDLDGFRKL FHRFLQEKGP SVDWGKIQRP PEDSIQPYEK IKARGLPDNI SSVLNKLVVV KLNGGLGTSM GCKGPKSLIG VRNENTFLDL TVQQIEHLNK TYNTDVPLVL MNSFNTDEDT KKILQKYNHC RVKIYTFNQS RYPRINKESL LPVAKDVSYS GENTEAWYPP GHGDIYASFY NSGLLDTFIG EGKEYIFVSN IDNLGATVDL YILNHLMNPP NGKRCEFVME VTNKTRADVK GGTLTQYEGK LRLVEIAQVP KAHVDEFKSV SKFKIFNTNN LWISLAAVKR LQEQNAIDME IIVNAKTLDG GLNVIQLETA VGAAIKSFEN SLGINVPRSR FLPVKTTSDL LLVMSNLYSL NAGSLTMSEK REFPTVPLVK LGSSFTKVQD YLRRFESIPD MLELDHLTVS GDVTFGKNVS LKGTVIIIAN HGDRIDIPPG AVLENKIVSG NLRILDH | |
Greater than 90% as determined by SDS-PAGE. |