missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcriptional coactivator YAP1 Protein
A DNA sequence encoding the Homo sapiens (Human) Transcriptional coactivator YAP1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9470-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001123617.1 | |
Transcriptional coactivator YAP1 Protein | |
Research Use Only | |
YAP1 | |
Recombinant Protein | |
Yeast | |
MDPGQQPPPQ PAPQGQGQPP SQPPQGQGPP SGPGQPAPAA TQAAPQAPPA GHQIVHVRGD SETDLEALFN AVMNPKTANV PQTVPMRLRK LPDSFFKPPE PKSHSRQAST DAGTAGALTP QHVRAHSSPA SLQLGAVSPG TLTPTGVVSG PAATPTAQHL RQSSFEIPDD VPLPAGWEMA KTSSGQRYFL NHIDQTTTWQ DPRKAMLSQM NVTAPTSPPV QQNMMNSASG PLPDGWEQAM TQDGEIYYIN HKNKTTSWLD PRLDPRFAMN QRISQSAPVK QPPPLAPQSP QGGVMGGSNS NQQQQMRLQQ LQMEKERLRL KQQELLRQAM RNINPSTANS PKCQELALRS QLPTLEQDGG TQNPVSSPGM SQELRTMTTN SSDPFLNSGT YHSRDESTDS GLSMSSYSVP RTPDDFLNSV DEMDTGDTIN QSTLPSQQNR FPDYLEAIPG TNVDLGTLEG DGMNIEGEEL MPSLQEALSS DILNDMESVL AATKLDKESF LTWL | |
Greater than 90% as determined by SDS-PAGE. |
10413 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |