missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcriptional coactivator YAP1 Protein
A DNA sequence encoding the Homo sapiens (Human) Transcriptional coactivator YAP1, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Accession Number | NP_001123617.1 |
---|---|
Gene ID (Entrez) | 10413 |
Name | Transcriptional coactivator YAP1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16098371
|
enQuireBio™
QP9470-YE-100UG |
100 μg |
645.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038381
|
enQuireBio™
QP9470-YE-500UG |
500 μg |
1740.00 €
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16008381
|
enQuireBio™
QP9470-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028381
|
enQuireBio™
QP9470-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16048381
|
enQuireBio™
QP9470-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018381
|
enQuireBio™
QP9470-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001123617.1 | |
Transcriptional coactivator YAP1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
10413 | |
Research Use Only | |
YAP1 | |
Recombinant Protein | |
Yeast | |
MDPGQQPPPQ PAPQGQGQPP SQPPQGQGPP SGPGQPAPAA TQAAPQAPPA GHQIVHVRGD SETDLEALFN AVMNPKTANV PQTVPMRLRK LPDSFFKPPE PKSHSRQAST DAGTAGALTP QHVRAHSSPA SLQLGAVSPG TLTPTGVVSG PAATPTAQHL RQSSFEIPDD VPLPAGWEMA KTSSGQRYFL NHIDQTTTWQ DPRKAMLSQM NVTAPTSPPV QQNMMNSASG PLPDGWEQAM TQDGEIYYIN HKNKTTSWLD PRLDPRFAMN QRISQSAPVK QPPPLAPQSP QGGVMGGSNS NQQQQMRLQQ LQMEKERLRL KQQELLRQAM RNINPSTANS PKCQELALRS QLPTLEQDGG TQNPVSSPGM SQELRTMTTN SSDPFLNSGT YHSRDESTDS GLSMSSYSVP RTPDDFLNSV DEMDTGDTIN QSTLPSQQNR FPDYLEAIPG TNVDLGTLEG DGMNIEGEEL MPSLQEALSS DILNDMESVL AATKLDKESF LTWL | |
Greater than 90% as determined by SDS-PAGE. |