missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transaldolase Protein

A DNA sequence encoding the Homo sapiens (Human) Transaldolase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8473-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16084801

  • 530.00 € / 100µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006746.1
Transaldolase Protein
Research Use Only
TAL
Recombinant Protein
E. coli
QRMESALDQL KQFTTVVADT GDFHAIDEYK PQDATTNPSL ILAAAQMPAY QELVEEAIAY GRKLGGSQED QIKNAIDKLF VLFGAEILKK IPGRVSTEVD ARLSFDKDAM VARARRLIEL YKEAGISKDR ILIKLSSTWE GIQAGKELEE QHGIHCNMTL LFSFAMS-QA VACAEAGVTL ISPFVGRILD WHVANTDKKS YEPLEDPGVK SVTKIYNYYK KFSYKTIVMG ASFRNTGEIK ALAGCDFLTI SPKLLGELLQ DNAKLVPVLS AKAAQASDLE KIHLDEKSFR WLHNEDQMAV EKLSDGIRKF AADAVKLERM LTERMFNAEN GK
Greater than 90% as determined by SDS-PAGE.
6888
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions