missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TOMM40, homolog Protein
A DNA sequence encoding the Homo sapiens (Human) TOMM40, homolog, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6821-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001122388.1 | |
TOMM40, homolog Protein | |
Research Use Only | |
TOMM40 | |
Recombinant Protein | |
E. coli | |
MGNVLAASSP PAGPPPPPAP ALVGLPPPPP SPPGFTLPPL GGSLGAGTST SRSSERTPGA ATASASGAAE DGACGCLPNP GTFEECHRKC KELFPIQMEG VKLTVNKGLS NHFQVNHTVA LSTIGESNYH FGVTYVGTKQ LSPTEAFPVL VGDMDNSGSL NAQVIHQLGP GLRSKMAIQT QQSKFVNWQV DGEYRGSDFT AAVTLGNPDV LVGSGILVAH YLQSITPCLA LGGELVYHRR PGEEGTVMSL AGKYTLNNWL ATVTLGQAGM HATYYHKASD QLQVGVEFEA STRMQDTSVS FGYQLDLPKA NLLFKGSVDS NWIVGATLEK KLPPLPLTLA LGAFLNHRKN KFQCGFGLTI G | |
Greater than 80% as determined by SDS-PAGE. |
10452 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |