missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TOMM40, homolog Protein
A DNA sequence encoding the Homo sapiens (Human) TOMM40, homolog, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_001122388.1 |
---|---|
Gene ID (Entrez) | 10452 |
Name | TOMM40, homolog Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16076070
|
enQuireBio™
QP6821-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16096070
|
enQuireBio™
QP6821-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16086070
|
enQuireBio™
QP6821-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16006080
|
enQuireBio™
QP6821-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016080
|
enQuireBio™
QP6821-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026080
|
enQuireBio™
QP6821-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001122388.1 | |
TOMM40, homolog Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
10452 | |
Research Use Only | |
TOMM40 | |
Recombinant Protein | |
E. coli | |
MGNVLAASSP PAGPPPPPAP ALVGLPPPPP SPPGFTLPPL GGSLGAGTST SRSSERTPGA ATASASGAAE DGACGCLPNP GTFEECHRKC KELFPIQMEG VKLTVNKGLS NHFQVNHTVA LSTIGESNYH FGVTYVGTKQ LSPTEAFPVL VGDMDNSGSL NAQVIHQLGP GLRSKMAIQT QQSKFVNWQV DGEYRGSDFT AAVTLGNPDV LVGSGILVAH YLQSITPCLA LGGELVYHRR PGEEGTVMSL AGKYTLNNWL ATVTLGQAGM HATYYHKASD QLQVGVEFEA STRMQDTSVS FGYQLDLPKA NLLFKGSVDS NWIVGATLEK KLPPLPLTLA LGAFLNHRKN KFQCGFGLTI G | |
Greater than 80% as determined by SDS-PAGE. |