missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFRSF25 / DR3 / TNFRSF12 Protein
A DNA sequence encoding the Homo sapiens (Human) TNFRSF25 / DR3 / TNFRSF12, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7891-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001034753.1 | |
TNFRSF25 / DR3 / TNFRSF12 Protein | |
Research Use Only | |
TNFRSF25 | |
Recombinant Protein | |
E. coli | |
QGGTRSPRCD CAGDFHKKIG LFCCRGCPAG HYLKAPCTEP CGNSTCLVCP QDTFLAWENH HNSECARCQA CDEQASQVAL ENCSAVADTR CGCKPGWFVE CQVSQCVSSS PFYCQPCLDC GALHRHTRLL CSRRDTDCGT CLPGFYEHGD GCVSCPTSTL GSCPERCAAV CGWRQ | |
Greater than 90% as determined by SDS-PAGE. |
8718 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |