missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TNFRSF25 / DR3 / TNFRSF12 Protein

A DNA sequence encoding the Homo sapiens (Human) TNFRSF25 / DR3 / TNFRSF12, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7891-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16001431

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001034753.1
TNFRSF25 / DR3 / TNFRSF12 Protein
Research Use Only
TNFRSF25
Recombinant Protein
E. coli
QGGTRSPRCD CAGDFHKKIG LFCCRGCPAG HYLKAPCTEP CGNSTCLVCP QDTFLAWENH HNSECARCQA CDEQASQVAL ENCSAVADTR CGCKPGWFVE CQVSQCVSSS PFYCQPCLDC GALHRHTRLL CSRRDTDCGT CLPGFYEHGD GCVSCPTSTL GSCPERCAAV CGWRQ
Greater than 90% as determined by SDS-PAGE.
8718
200 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions