missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thioredoxin domain-containing protein 12 Protein
A DNA sequence encoding the Homo sapiens (Human) Thioredoxin domain-containing protein 12, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6857-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_056997.1 | |
Thioredoxin domain-containing protein 12 Protein | |
Research Use Only | |
TXNDC12 | |
Recombinant Protein | |
E. coli | |
HNGLGKGFGD HIHWRTLEDG KKEAAASGLP LMVIIHKSWC GACKALKPKF AESTEISELS HNFVMVNLED EEEPKDEDFS PDGGYIPRIL FLDPSGKVHP EIINENGNPS YKYFYVSAEQ VVQGMKEAQE RLTGDAFRKK HLEDEL | |
Greater than 90% as determined by SDS-PAGE. |
51060 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |