missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TALLA-1 / TSPAN7 Protein
A DNA sequence encoding the Homo sapiens (Human) TALLA-1 / TSPAN7, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Accession Number | NP_004606.2 |
---|---|
Gene ID (Entrez) | 7102 |
Name | TALLA-1 / TSPAN7 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16078331
|
enQuireBio™
QP9449-YE-100UG |
100 μg |
645.00 €
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018341
|
enQuireBio™
QP9449-YE-500UG |
500 μg |
1740.00 €
500µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088331
|
enQuireBio™
QP9449-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008341
|
enQuireBio™
QP9449-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028341
|
enQuireBio™
QP9449-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098331
|
enQuireBio™
QP9449-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004606.2 | |
TALLA-1 / TSPAN7 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
7102 | |
Research Use Only | |
TSPAN7 | |
Recombinant Protein | |
Yeast | |
RHEIKDTFLR TYTDAMQTYN GNDERSRAVD HVQRSLSCCG VQNYTNWSTS PYFLEHGIPP SCCMNETDCN PQDLHNLTVA ATKVNQKGCY DLVTSFMETN MGIIAGVAFG I | |
Greater than 90% as determined by SDS-PAGE. |